Invitrogen
Manufacturer Code:PA568039
Catalog # PIPA568039
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthesized peptide derived from human INSL5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: insulin-like 5; Insulin-like peptide 5; Insulin-like peptide INSL5; Insulin-like peptide INSL5 A chain; Insulin-like peptide INSL5 B chain; prepro-INSL5; Relaxin/insulin-like factor 2; Relaxin/insulin-like protein
Gene Aliases: INSL5; PRO182; Rif; RIF2; UNQ156; UNQ156/PRO182; Zins3
UniProt ID: (Human) Q9Y5Q6, (Mouse) Q9WUG6
Entrez Gene ID: (Human) 10022, (Mouse) 23919
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.