Invitrogen
Manufacturer Code:PA541707
Catalog # PIPA541707
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human INSIG2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: INSIG-2; INSIG-2 membrane protein; INSIG2 membrane protein; insulin induced factor 2; Insulin induced gene 2 MGC26273; insulin induced protein 2; insulin-induced 2 protein; Insulin-induced gene 2 protein; insulin-induced membrane protein 2
Gene Aliases: 2900053I11Rik; C730043J18Rik; INSIG-2; INSIG2
UniProt ID: (Human) Q9Y5U4, (Pig) Q6PQZ3, (Rat) Q80UA9, (Mouse) Q91WG1
Entrez Gene ID: (Rabbit) 100355934, (Sheep) 101110569, (Human) 51141, (Goat) 100861144, (Dog) 610496, (Pig) 100170127, (Horse) 100049957, (Rat) 288985, (Mouse) 72999
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.