Invitrogen
Manufacturer Code:PA5109863
Catalog # PIPA5109863
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 50-150 of human INSIG2 (NP_057217.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2900053I11Rik BOS_1878 C730043J18Rik Insig2 INSIG-2 Insulin-induced gene 2 protein MGC26273; INSIG-2; INSIG-2 membrane protein; INSIG2 membrane protein; insulin induced protein 2; Insulin-induced gene 2 protein
Gene Aliases: 2900053I11Rik; C730043J18Rik; INSIG-2; INSIG2
UniProt ID: (Human) Q9Y5U4, (Mouse) Q91WG1
Entrez Gene ID: (Human) 51141, (Mouse) 72999
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.