Invitrogen
Manufacturer Code:PA541706
Catalog # PIPA541706
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human INSIG1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: growth response protein (CL-6); Immediate-early protein CL-6; INSIG-1; INSIG-1 membrane protein; Insulin induced gene 1 CL-6 MGC1405 CL6; insulin induced protein 1; Insulin-induced gene 1 protein; Insulin-induced growth response protein CL-6
Gene Aliases: 1810013C12Rik; CL-6; CL6; fb55a03; fi35b10; Insig-1; INSIG1; wu:fb55a03; wu:fi35b10; zgc:55439
UniProt ID: (Human) O15503, (Bovine) A0JNC3, (Mouse) Q8BGI3, (Rat) Q08755, (Zebrafish) Q8AV61
Entrez Gene ID: (Human) 3638, (Sheep) 101106422, (Dog) 475554, (Bovine) 511899, (Horse) 100065774, (Rabbit) 100353280, (Goat) 102183937, (Mouse) 231070, (Guinea pig) 100718114, (Rat) 64194, (Zebrafish) 334189
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.