Novus Biologicals
Manufacturer Code:NB11055244AF750
Catalog # N110244A70
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide representing an internal region of the human INSIG-1 protein (between residues 1-100) according to Swiss-ProtNo. O15503. |
Conjugate | Alexa Fluor 750 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
Protein Aliases: CL6 CL-6 INSIG-1 INSIG-1 membrane protein insulin induced gene 1 insulin-induced gene 1 protein MGC1405; INSIG-1; INSIG-1 membrane protein; Insulin-induced gene 1 protein
Gene Aliases: CL-6; CL6; INSIG1
UniProt ID: (Human) O15503
Entrez Gene ID: (Human) 3638
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.