Abnova
Manufacturer Code:RAB00497
Catalog # 89-337-922
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | rIRDN/805 |
Immunogen | Original antibody is raised against recombinant protein corresponding to full length human INS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C |
Mouse Monoclonal Antibody.
Protein Aliases: ILPR IRDN; Insulin; Insulin A chain; Insulin B chain; preproinsulin; proinsulin
Gene Aliases: IDDM; IDDM1; IDDM2; ILPR; INS; IRDN; MODY10
UniProt ID: (Human) P01308
Entrez Gene ID: (Human) 3630
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.