Abnova
Manufacturer Code:H00051763A01
Catalog # 89-106-932
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 147 a.a. &bsim 253 a.a) partial recombinant protein with GST tag.,INPP5K (NP_057616 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant INPP5K.
For Research Use Only
Protein Aliases: Inositol polyphosphate 5-phosphatase K; Phosphatidylinositol-3,4,5-trisphosphate 5-phosphatase; Phosphatidylinositol-4,5-bisphosphate 5-phosphatase; PPS; Skeletal muscle and kidney-enriched inositol phosphatase; SKIP
Gene Aliases: INPP5K; PPS; SKIP
UniProt ID: (Human) Q9BT40
Entrez Gene ID: (Human) 51763
Molecular Function:
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.