Proteintech
Manufacturer Code:150981AP150UL
Catalog # 50-172-9976
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | INPP5K Fusion Protein Ag7192 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C62; Inositol polyphosphate 5-phosphatase K; INPP5K PPS SKIP; Phosphatidylinositol-3,4,5-trisphosphate 5-phosphatase; Phosphatidylinositol-4,5-bisphosphate 5-phosphatase; PI-5-phosphatase related; putative phosphoinositide 5-phosphatase type II; putative phosphoinositide 5-phosphatase type II; C62; putative PI-5-phosphatase; skeletal muscle and kidney enriched inositol phosphatase; skeletal muscle and kidney enriched inositol polyphosphate phosphatase; Skeletal muscle and kidney-enriched inositol phosphatase
Gene Aliases: C62; INPP5K; PPS; SKIP
UniProt ID: (Human) Q9BT40, (Mouse) Q8C5L6
Entrez Gene ID: (Human) 51763, (Rat) 287533, (Mouse) 19062
Molecular Function:
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.