Invitrogen
Manufacturer Code:PA532876
Catalog # PIPA532876
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 15 amino acid peptide from N-terminus of human INPP5J / PIB5PA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Inositol polyphosphate 5-phosphatase J; INPP5 PIB5PA PIPP inositol polyphosphate 5-phosphatase J phosphatidylinositol (45) bisphosphate 5-phosphatase A phosphatidylinositol 45-bisphosphate 5-phosphatase A; phosphatidylinositol (4,5) bisphosphate 5-phosphatase, A; Phosphatidylinositol 4,5-bisphosphate 5-phosphatase A
Gene Aliases: INPP5; INPP5J; PIB5PA; PIPP
UniProt ID: (Human) Q15735
Entrez Gene ID: (Human) 27124
Molecular Function:
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.