Invitrogen
Manufacturer Code:MA527229
Catalog # PIMA527229
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1D4 |
Immunogen | Full length human recombit protein of INPP5F produced in HEK293T cell |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: hSAC2; Inositol polyphosphate 5-phosphatase F; MSTP007 MSTPO47 SAC2 hSAC2 Sac domain-containing inositol phosphatase 2 phosphatidylinositide phosphatase SAC2 sac domain-containing phosphoinositide 5-phosphatase 2; Phosphatidylinositide phosphatase SAC2; Sac domain-containing inositol phosphatase 2; Sac domain-containing phosphoinositide 4-phosphatase 2; sac domain-containing phosphoinositide 5-phosphatase 2
Gene Aliases: hSAC2; INPP5F; KIAA0966; MSTP007; MSTP047; MSTPO47; SAC2
UniProt ID: (Human) Q9Y2H2
Entrez Gene ID: (Human) 22876
Molecular Function:
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.