Novus Biologicals
Manufacturer Code:NBP2840930.1ML
Catalog # NB032021
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of Human INPP5F |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.1.3 EC 3.1.3.- EC 3.1.3.16 EC 3.1.3.36 FLJ13081 hSac2 Inositol polyphosphate 5-phosphatase F inositol polyphosphate-5-phosphatase F KIAA0966MGC59773 MGC131851 phosphatidylinositide phosphatase SAC2 Sac domain-containing inositol phosphatase 2 Sac domain-containing phosphoinositide 5-phosphatase 2 SAC2MSTPO47; hSAC2; Inositol polyphosphate 5-phosphatase F; Phosphatidylinositide phosphatase SAC2; Sac domain-containing inositol phosphatase 2; Sac domain-containing phosphoinositide 4-phosphatase 2; sac domain-containing phosphoinositide 5-phosphatase 2
Gene Aliases: hSAC2; INPP5F; KIAA0966; MSTP007; MSTP047; MSTPO47; SAC2
UniProt ID: (Human) Q9Y2H2
Entrez Gene ID: (Human) 22876
Molecular Function:
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.