Invitrogen
Manufacturer Code:PA523992
Catalog # PIPA523992
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 950-979 amino acids from the C-terminal region of human INPP5B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5PTase; 5PTase 75 kDa inositol polyphosphate-5-phosphatase phosphoinositide 5-phosphatase type II inositol-145-trisphosphate 5-phosphatase; 75 kDa inositol polyphosphate-5-phosphatase; inositol polyphosphate-5-phosphatase, 75kDa; Phosphoinositide 5-phosphatase; Type II inositol 1,4,5-trisphosphate 5-phosphatase
Gene Aliases: 5PTase; INPP5B; OCRL2
UniProt ID: (Human) P32019
Entrez Gene ID: (Human) 3633
Molecular Function: hydrolase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.