Invitrogen
Manufacturer Code:PA552921
Catalog # PIPA552921
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human INPP5A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 43 kDa inositol polyphosphate 5-phophatase; 43 kDa inositol polyphosphate 5-phosphatase; 5PTase; 5PTASE 43 kDa inositol polyphosphate 5-phophatase CTCL tumor antigen HD-CL-02 InsP3 5-phosphatase inositol trisphosphate-5-phosphatase 40kD type I inositol-145-trisphosphate 5-phophatase type I inositol-145-trisphosphate 5-phosphatase; CTCL tumor antigen HD-CL-02; Inositol polyphosphate-5-phosphatase A; inositol polyphosphate-5-phosphatase, 40kD; inositol polyphosphate-5-phosphatase, 40kDa; inositol trisphosphate-5-phosphatase, 40kD; InsP3 5-phosphatase; Type I inositol 1,4,5-trisphosphate 5-phosphatase; type I inositol-1,4,5-trisphosphate 5-phophatase; type I inositol-1,4,5-trisphosphate 5-phosphatase
Gene Aliases: 5PTASE; INPP5A
UniProt ID: (Human) Q14642
Entrez Gene ID: (Human) 3632
Molecular Function:
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.