Invitrogen
Manufacturer Code:PA558057
Catalog # PIPA558057
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human INPP4B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: inositol polyphosphate 4-phosphatase II; 4-phosphatase II; Inositol polyphosphate 4-phosphatase type II; inositol polyphosphate-4-phosphatase, type II, 105kD; inositol polyphosphate-4-phosphatase, type II, 105kDa; Type II inositol 3 4-bisphosphate 4-phosphatase Inositol polyphosphate 4-phosphatase type II INPP4B; Type II inositol 3,4-bisphosphate 4-phosphatase; type II inositol-3,4-bisphosphate 4-phosphatase
Gene Aliases: INPP4B
UniProt ID: (Human) O15327
Entrez Gene ID: (Human) 8821
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.