Invitrogen
Manufacturer Code:PA531354
Catalog # PIPA531354
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 1 and 228 of Human INMT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Amine N-methyltransferase; amine N-methyltransferase aromatic alkylamine N-methyltransferase arylamine N-methyltransferase indolamine N-methyltransferase nicotine N-methyltransferase; Aromatic alkylamine N-methyltransferase; arylamine N-methyltransferase; Indolamine N-methyltransferase; Indolethylamine N-methyltransferase; nicotine N-methyltransferase; TEMT; Thioether S-methyltransferase
Gene Aliases: INMT; TEMT
UniProt ID: (Human) O95050
Entrez Gene ID: (Human) 11185
Molecular Function:
methyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.