Proteintech
Manufacturer Code:215781AP150UL
Catalog # 50-173-3497
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | INMT Fusion Protein Ag16121 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Amine N methyltransferase Arylamine N methyltransferase Indolamine N methyltransferase INMT; Amine N-methyltransferase; Aromatic alkylamine N-methyltransferase; arylamine N-methyltransferase; Indolamine N-methyltransferase; Indolethylamine N-methyltransferase; nicotine N-methyltransferase; TEMT; Thioether S-methyltransferase
Gene Aliases: INMT; TEMT
UniProt ID: (Human) O95050, (Mouse) P40936
Entrez Gene ID: (Human) 11185, (Mouse) 21743
Molecular Function:
methyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.