Novus Biologicals
Manufacturer Code:NBP290261AF647
Catalog # NB038351
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 119 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Human ING5 (Q8WYH8-2 Glu36-Gln84). |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: inhibitor of growth family member 5 inhibitor of growth protein 5 p28ING5FLJ23842; inhibitor of growth family, member 5; Inhibitor of growth protein 5; p28ING5
Gene Aliases: ING5; p28ING5
UniProt ID: (Human) Q8WYH8
Entrez Gene ID: (Human) 84289
Molecular Function: DNA binding protein chromatin/chromatin-binding protein nucleic acid binding transcription cofactor transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.