Proteintech
Manufacturer Code:106651AP150UL
Catalog # 50-172-6493
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ING5 Fusion Protein Ag1059 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ING5 Inhibitor of growth protein 5 p28ING5; inhibitor of growth family, member 5; Inhibitor of growth protein 5; p28ING5
Gene Aliases: 1700001C14Rik; 1700027H23Rik; 1810018M11Rik; AI225768; ING5; p28ING5
UniProt ID: (Human) Q8WYH8, (Mouse) Q9D8Y8
Entrez Gene ID: (Human) 84289, (Rat) 363292, (Mouse) 66262
Molecular Function:
DNA binding protein
chromatin/chromatin-binding protein
nucleic acid binding
transcription cofactor
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.