Proteintech
Manufacturer Code:161881AP150UL
Catalog # 50-173-0779
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: brain my036 protein; candidate tumor suppressor p33 ING1 homolog; ING1-like protein; ING4 Inhibitor of growth protein 4 my036 p29ING4; Inhibitor of growth protein 4; p29ING4
Gene Aliases: D6Wsu147e; D6Xrf92; ING4; My036; p29ING4
UniProt ID: (Human) Q0EF63, (Mouse) Q8C0D7
Entrez Gene ID: (Human) 51147, (Rat) 297597, (Mouse) 28019
Molecular Function:
DNA binding protein
chromatin/chromatin-binding protein
nucleic acid binding
transcription cofactor
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.