Proteintech
Manufacturer Code:665121IG150UL
Catalog # 50-173-6934
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2C3A7 |
Immunogen | ING2 Fusion Protein Ag19291 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ING1L ING1Lp ING2 Inhibitor of growth protein 2 p32 p33ING2; ING1Lp; Inhibitor of growth 1-like protein; inhibitor of growth family, member 2; Inhibitor of growth protein 2; p32; p33ING2
Gene Aliases: 2810011M06Rik; ING1L; ING2; Ing2b; P33ING2
UniProt ID: (Human) Q9H160, (Mouse) Q9ESK4
Entrez Gene ID: (Human) 3622, (Mouse) 69260, (Rat) 290744
Molecular Function:
DNA binding protein
chromatin/chromatin-binding protein
nucleic acid binding
transcription cofactor
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.