Proteintech
Manufacturer Code:204661AP150UL
Catalog # 50-173-2908
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | INF2 Fusion Protein Ag14302 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C14orf151 C14orf173 HBEBP2 binding protein C INF2 Inverted formin 2 pp9484; formin, inverted; HBEAG-binding protein 2 binding protein C; HBEBP2-binding protein C; Inverted formin-2
Gene Aliases: 2610204M08Rik; AA589465; AW125550; C14orf151; C14orf173; CMTDIE; EG629699; FSGS5; INF2; pp9484
UniProt ID: (Human) Q27J81, (Mouse) Q0GNC1
Entrez Gene ID: (Human) 64423, (Mouse) 70435
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.