Invitrogen
Manufacturer Code:PA5102742
Catalog # PIPA5102742
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human IMPDH2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HAMAP-Rule:MF_03156} IMP (inosine 5'-monophosphate) dehydrogenase 2 IMP (inosine monophosphate) dehydrogenase 2 IMP dehydrogenase 2 IMP dehydrogenase 2 {ECO:0000255 IMP dehydrogenase type II IMP oxireductase 2 IMPD IMPD 2 IMPD 2 {ECO:0000255 IMPD2 IMPDH 2 IMPDH 2 {ECO:0000255 IMPDH II IMPDH2 IMPDH-II inosine 5' phosphate dehydrogenase 2 inosine 5-monophosphate dehydrogenase 2 inosine 5'-phosphate dehydrogenase 2 inosine monophosphate dehydrogenase 2 inosine monophosphate dehydrogenase type II inosine-5'-monophosphate dehydrogenase 2 inosine-5'-monophosphate dehydrogenase 2 {ECO:0000255; IMP (inosine 5'-monophosphate) dehydrogenase 2; IMP (inosine monophosphate) dehydrogenase 2; IMP dehydrogenase 2; IMP dehydrogenase 2 {ECO:0000255|HAMAP-Rule:MF_03156}; IMP dehydrogenase type II; IMP oxireductase 2; IMPD 2; IMPD 2 {ECO:0000255|HAMAP-Rule:MF_03156}; IMPDH 2; IMPDH 2 {ECO:0000255|HAMAP-Rule:MF_03156}; IMPDH-II; IMPDH2; inosine 5' phosphate dehydrogenase 2; inosine 5'-phosphate dehydrogenase 2; inosine 5-monophosphate dehydrogenase 2; inosine monophosphate dehydrogenase 2; inosine monophosphate dehydrogenase type II; Inosine-5'-monophosphate dehydrogenase 2; inosine-5'-monophosphate dehydrogenase 2 {ECO:0000255|HAMAP-Rule:MF_03156}
Gene Aliases: IMPD; IMPD 2; IMPD2; IMPDH 2; IMPDH-II; IMPDH2
UniProt ID: (Human) P12268, (Mouse) P24547
Entrez Gene ID: (Human) 3615, (Rat) 301005, (Mouse) 23918
Molecular Function: dehydrogenase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.