Abnova
Manufacturer Code:PAB19702
Catalog # 89-121-229
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to internal region of human IMPDH2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of IMPDH2.
For Research Use Only
Protein Aliases: IMP (inosine 5'-monophosphate) dehydrogenase 2; IMP (inosine monophosphate) dehydrogenase 2; IMP dehydrogenase 2; IMP oxireductase 2; IMPD 2; IMPD2; IMPDH 2; IMPDH-II; IMPDH2; inosine 5' phosphate dehydrogenase 2; inosine monophosphate dehydrogenase type II; Inosine-5'-monophosphate dehydrogenase 2
Gene Aliases: IMPD2; IMPDH-II; IMPDH2
UniProt ID: (Human) P12268
Entrez Gene ID: (Human) 3615
Molecular Function: dehydrogenase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.