Invitrogen
Manufacturer Code:PA547893
Catalog # PIPA547893
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | CHO-derived recombit mouse Inositol Monophosphatase 3/IMPAD1 Asn145-His356 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: 3'(2'), 5'-bisphosphate nucleotidase 2; Ac2-190; Golgi 3-prime phosphoadenosine 5-prime phosphate 3-prime phosphatase; Golgi-resident adenosine 3',5'-bisphosphate 3'-phosphatase; Golgi-resident PAP phosphatase; IMP 3; IMP3 IMPA3 IMPase 3 EC 3.1.3 EC 3.1.3.25 FLJ20421 IMP 3 IMPA3IMPase 3 inositol monophosphatase 3 inositol monophosphatase domain containing 1 Inositol monophosphatase domain-containing protein 1 Inositol-1(or 4)-monophosphatase 3 Myo-inositol monophosphatase A3; IMPase 3; inositol monophosphatase 3; Inositol monophosphatase domain-containing protein 1; inositol-1(or 4)-monophosphatase 3; Myo-inositol monophosphatase A3; Phosphoadenosine phosphate 3'-nucleotidase
Gene Aliases: 1110001C20Rik; AA408880; AI451589; AL022796; B230207P20; Bpnt2; gPAPP; IMP 3; Impa3; Impad1; Jaws; RGD1306455
UniProt ID: (Mouse) Q80V26, (Rat) D4AD37
Entrez Gene ID: (Mouse) 242291, (Rat) 312952
Molecular Function:
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.