Novus Biologicals
Manufacturer Code:NBP297107AF405
Catalog # NB081084
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant IMPA1 (P29218-1 Met1-Asp277). |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D-galactose 1-phosphate phosphatase; EC 3.1.3.25 IMP IMP 1 IMPA IMPase 1 inositol monophosphatase 1 inositol(myo)-1(or 4)-monophosphatase 1 Inositol-1(or 4)-monophosphatase 1 Lithium-sensitive myo-inositol monophosphatase A1; IMP 1; IMPase 1; Inositol monophosphatase 1; inositol(myo)-1(or 4)-monophosphatase 1; Inositol-1(or 4)-monophosphatase 1; Lithium-sensitive myo-inositol monophosphatase A1; myo-inositol monophosphatase 1; testicular tissue protein Li 94
Gene Aliases: IMP; IMPA; IMPA1
UniProt ID: (Human) P29218
Entrez Gene ID: (Human) 3612
Molecular Function:
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.