Invitrogen
Manufacturer Code:PA531673
Catalog # PIPA531673
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 184 of Human IMP3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BRMS2; BRMS2 C15orf12 MRPS4 U3 small nucleolar ribonucleoprotein protein IMP3 U3 snoRNP protein 3 homolog U3 snoRNP protein IMP3; IMP3, U3 small nucleolar ribonucleoprotein, homolog; U3 small nucleolar ribonucleoprotein protein IMP3; U3 snoRNP protein 3 homolog; U3 snoRNP protein IMP3
Gene Aliases: BRMS2; C15orf12; IMP3; MRPS4
UniProt ID: (Human) Q9NV31
Entrez Gene ID: (Human) 55272
Molecular Function:
RNA binding protein
nucleic acid binding
ribonucleoprotein
ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.