R&D Systems
Manufacturer Code:MAB2078RSP
Catalog # MAB2078RSP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 287219R |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human LILRB2/CD85d/ILT4 Gly24-His458 Accession # ACT64556 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70°C as supplied. 1 month 2 to 8°C under sterile conditions after reconstitution. 6 months -20 to -70°C under sterile conditions a |
Mouse Monoclonal Antibody
Protein Aliases: CD85 antigen-like family member D; CD85 antigen-like family member D CD85d CD85d antigen Ig-like transcript 4 ILT4 ILT-4 ILT4CD85d Immunoglobulin-like transcript 4 Leukocyte immunoglobulin-like receptor 2 leukocyte immunoglobulin-like receptor subfamily B member 2 leukocyte immunoglobulin-like receptor subfamily B member 2 soluble isoform leukocyte immunoglobulin-like receptor subfamily B (with TM and ITIM domains) LIR2 LIR2CD85D LIR-2subfamily A (with TM domain) member 6 member 2 MIR10 MIR10LILRA6 Monocyte/macrophage immunoglobulin-like receptor 10; Ig-like transcript 4; leucocyte Ig-like receptor B2; leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 2; monocyte/macrophage immunoglobulin-like receptor 10; myeloid inhibitory receptor 10
Gene Aliases: CD85D; ILT-4; ILT4; LIR-2; LIR2; MIR-10; MIR10
Entrez Gene ID: (Human) 10288
Molecular Function:
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
membrane-bound signaling molecule
receptor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.