R&D Systems
Manufacturer Code:FAB20171F025
Catalog # FAB20171F02
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 292305 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human ILT2/CD85j Gly24-His458 Accession # Q8NHL6 |
Conjugate | Fluorescein |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD85 antigen-like family member J; CD85 antigen-like family member J CD85J CD85j antigen Ig-like transcript 2 ILT2 ILT-2 ILT2FLJ37515 Immunoglobulin-like transcript 2 leukocyte Ig-like receptor-1 Leukocyte immunoglobulin-like receptor 1 leukocyte immunoglobulin-like receptor subfamily B member 1 leukocyte immunoglobulin-like receptor subfamily B member 1 soluble isoform leukocyte immunoglobulin-like receptor subfamily B (with TM and ITIM domains) LIR1 LIR1LIR-1MIR-7CD85MIR7CD85 antigen member 1 MIR7 Monocyte/macrophage immunoglobulin-like receptor 7; CD85j; Ig-like transcript 2; ILT-2; Immunoglobulin-like transcript 2; leucocyte Ig-like receptor B1; Leukocyte immunoglobulin-like receptor subfamily B member 1; leukocyte immunoglobulin-like receptor subfamily B member 1 soluble isoform; leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 1; LIR-1; MIR-7; Monocyte/macrophage immunoglobulin-like receptor 7; myeloid inhibitory receptor 7
Gene Aliases: CD85J; ILT-2; ILT2; LILRB1; LIR-1; LIR1; MIR-7; MIR7; PIR-B; PIRB
UniProt ID: (Human) Q8NHL6
Entrez Gene ID: (Human) 10859
Molecular Function:
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
membrane-bound signaling molecule
receptor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.