R&D Systems
Manufacturer Code:MAB4266
Catalog # MAB4266
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 443208 |
Immunogen | <EM>E. coli</EM>-derived recombinant human ILK Met1-Lys452 Accession # Q13418 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 59 kDa serine/threonine-protein kinase; 59 kDa serine/threonine-protein kinase DKFZp686F1765 EC 2.7.11.1 ESTM24 ILK1 ILK-1 ILK2 ILK-2 integrin-linked kinase integrin-linked kinase-2 integrin-linked protein kinase p59 p59ILK; Beta-integrin-linked kinase; epididymis secretory protein Li 28; ILK-1; ILK-2; integrin-linked kinase; integrin-linked kinase-2; Integrin-linked protein kinase; p59ILK
Gene Aliases: HEL-S-28; ILK; ILK-1; ILK-2; ILK1; ILK2; P59; p59ILK
UniProt ID: (Human) Q13418
Entrez Gene ID: (Human) 3611
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.