Novus Biologicals
Manufacturer Code:NBP237448
Catalog # NBP237448
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3A9 |
Immunogen | Purified recombinant fragment of human ILK (AA: 97-244) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 59 kDa serine/threonine-protein kinase; 59 kDa serine/threonine-protein kinase DKFZp686F1765 EC 2.7.11.1 ILK1 ILK-1 ILK2 ILK-2 integrin-linked kinase integrin-linked kinase-2 integrin-linked protein kinase P59 p59ILK; Beta-integrin-linked kinase; epididymis secretory protein Li 28; ILK-1; ILK-2; integrin-linked kinase; integrin-linked kinase-2; Integrin-linked protein kinase; p59ILK
Gene Aliases: HEL-S-28; ILK; ILK-1; ILK-2; ILK1; ILK2; P59; p59ILK
UniProt ID: (Human) Q13418
Entrez Gene ID: (Human) 3611
Molecular Function: kinase non-receptor serine/threonine protein kinase non-receptor tyrosine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.