Abnova
Manufacturer Code:PAB18513
Catalog # 89-120-075
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to residues surrounding S246 of human ILK. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of ILK.
For Research Use Only
Protein Aliases: 59 kDa serine/threonine-protein kinase; Beta-integrin-linked kinase; DKFZp686F1765; epididymis secretory protein Li 28; ILK-1; ILK-2; integrin-linked kinase; integrin-linked kinase-2; Integrin-linked protein kinase; P59; p59ILK
Gene Aliases: HEL-S-28; ILK; ILK-1; ILK-2; ILK1; ILK2; P59; p59ILK
UniProt ID: (Human) Q13418
Entrez Gene ID: (Human) 3611
Molecular Function: kinase non-receptor serine/threonine protein kinase non-receptor tyrosine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.