Novus Biologicals
Manufacturer Code:NBP122492AF532
Catalog # NP122492A32
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | RM0114-14F33 |
Immunogen | Mouse recombinant protein of the extracellular domain of IL-6 R |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rat Monoclonal Antibody
Protein Aliases: CD126; CD126 antigen; CD126 CD126 antigen gp80 IL-6 receptor subunit alpha IL-6R 1 IL-6R subunit alpha IL-6R-1 IL6RA IL-6RA IL-6R-alpha interleukin 6 receptor interleukin 6 receptor alpha subunit interleukin-6 receptor subunit alpha Membrane glycoprotein 80 MGC104991; gp80; IL-6 receptor subunit alpha; IL-6R 1; Interleukin-6 receptor subunit alpha; Membrane glycoprotein 80; sIL6R; Soluble interleukin-6 receptor subunit alpha
Gene Aliases: CD126; gp80; IL-6R-1; IL-6RA; IL6Q; IL6R; IL6RA; IL6RQ
UniProt ID: (Human) P08887
Entrez Gene ID: (Human) 3570
Molecular Function:
cytokine
defense/immunity protein
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.