Abnova
Manufacturer Code:H00003569M05
Catalog # 89-123-172
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3E8 |
Immunogen | IL6 (NP_000591 29 a.a.-212 a.a.) full-length recombinant protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant IL6.
For Research Use Only
Protein Aliases: B-cell differentiation factor; B-cell stimulatory factor 2; BSF-2; BSF2; CDF; CTL differentiation factor; HGF; HSF; Hybridoma growth factor; IFN-beta-2; IFNB2; IL-6; Interferon beta-2; interleukin BSF-2; Interleukin-6
Gene Aliases: BSF-2; BSF2; CDF; HGF; HSF; IFN-beta-2; IFNB2; IL-6; IL6
UniProt ID: (Human) P05231
Entrez Gene ID: (Human) 3569
Molecular Function:
cytokine
interleukin superfamily
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.