Invitrogen
Manufacturer Code:PA5115418
Catalog # PIPA5115418
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human IL4I1 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Fig-1 protein; FIG1 Fig-1 protein Fig1-ps hFIG1 Il4i1 IL4I1_2 IL4-induced protein 1 interleukin 4 induced 1 interleukin four induced 1 interleukin-4-induced protein 1 interleukin-four induced gene 1 interleukin-four induced gene-1 protein interleukin-four induced protein-1 LAAO L-amino acid oxidase L-amino-acid oxidase LAO LOC100360621 mFIG1 Protein Fig-1 rCG53598-like UNQ636/PRO1265; hFIG1; IL4-induced protein 1; interleukin four induced 1; Interleukin-4-induced protein 1; L-amino-acid oxidase; LAAO; Protein Fig-1; rCG53598-like
Gene Aliases: FIG1; IL4I1; LAAO; LAO; UNQ636/PRO1265
UniProt ID: (Human) Q96RQ9
Entrez Gene ID: (Human) 259307, (Rat) 100360621
Molecular Function: DNA binding protein DNA methyltransferase methyltransferase nucleic acid binding oxidase oxidoreductase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.