Proteintech
Manufacturer Code:131131AP150UL
Catalog # 50-172-8498
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | IL36RN Fusion Protein Ag3780 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FIL1 delta; FIL1 FIL1 delta FIL1(DELTA) FIL1D IL 1 delta IL 1 related protein 3 IL 1F5 IL 1HY1 IL 1L1 IL 1ra homolog 1 IL 1RP3 IL1F5 IL1HY1 IL1L1 IL1RP3 Interleukin 1 delta Interleukin 1 family member 5 Interleukin 1 HY1 Interleukin 1 like protein 1; IL-1 delta; IL-1 related protein 3; IL-1-related protein 3; IL-1F5; IL-1F5 (IL-1HY1, FIL1-delta, IL-1RP3, IL-1L1, IL-1-delta); IL-1HY1; IL-1L1; IL-1ra homolog 1; IL-1RP3; IL-36Ra; IL1F5 (Canonical product IL-1F5a); interleukin 1 family, member 5 (delta); Interleukin-1 delta; Interleukin-1 family member 5; Interleukin-1 HY1; Interleukin-1 receptor antagonist homolog 1; Interleukin-1-like protein 1; Interleukin-36 receptor antagonist protein
Gene Aliases: FIL1; FIL1(DELTA); FIL1D; IL-36Ra; IL1F5; IL1HY1; IL1L1; IL1RP3; IL36RA; IL36RN; PSORP; PSORS14; UNQ1896/PRO4342
UniProt ID: (Human) Q9UBH0
Entrez Gene ID: (Human) 26525
Molecular Function:
cytokine
interleukin superfamily
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.