Invitrogen
Manufacturer Code:PA5115413
Catalog # PIPA5115413
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human IL2RG corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: [g]c Ab2-183 CD132 CD132 antigen CIDX common cytokine receptor gamma chain cytokine receptor common subunit gamma gamma(c) gammaC gc IL-2 receptor subunit gamma IL2R gamma IL-2R subunit gamma IL2RG IL-2RG IMD4 interleukin 2 receptor subunit gamma interleukin 2 receptor gamma interleukin 2 receptor gamma (severe combined immunodeficiency) interleukin 2 receptor gamma chain Interleukin-2 receptor subunit gamma P64 SCIDX SCIDX1; CD132; CD132 antigen; common cytokine receptor gamma chain; Cytokine receptor common subunit gamma; gammaC; IL-2 receptor subunit gamma; IL-2R subunit gamma; interleukin 2 receptor, gamma; interleukin 2 receptor, gamma (severe combined immunodeficiency); Interleukin-2 receptor subunit gamma; p64
Gene Aliases: [g]c; Ab2-183; CD132; CIDX; gamma(c); gc; IL-2RG; IL2RG; IMD4; P64; SCIDX; SCIDX1
UniProt ID: (Human) P31785, (Mouse) P34902
Entrez Gene ID: (Human) 3561, (Rat) 140924, (Mouse) 16186
Molecular Function: cytokine defense/immunity protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.