Abnova
Manufacturer Code:PAB13096
Catalog # 89-116-348
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to N-terminus of human IL27RA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of IL27RA.
For Research Use Only
Protein Aliases: class I cytokine receptor; CRL1; Cytokine receptor WSX-1; Cytokine receptor-like 1; IL-27 receptor subunit alpha; IL-27R subunit alpha; IL-27R-alpha; IL27R; interleukin 27 receptor, alpha; Interleukin-27 receptor subunit alpha; T-cell cytokine receptor type 1; TCCR; Type I T-cell cytokine receptor; WSX1; ZcytoR1
Gene Aliases: CRL1; IL-27RA; IL27R; IL27RA; TCCR; UNQ296/PRO336; WSX1; zcytor1
UniProt ID: (Human) Q6UWB1
Entrez Gene ID: (Human) 9466
Molecular Function:
cytokine
defense/immunity protein
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.