Invitrogen
Manufacturer Code:MA524013
Catalog # PIMA524013
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 303308 |
Immunogen | E. coli-derived recombit mouse IL-24 Arg27-His181 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: C49A FISP IL10B MDA7 MOB5 ST16 IL-4-induced secreted protein interleukin-24 melanocyte-associated Mda-7 melanoma differentiation-associated gene 7 protein suppression of tumorigenicity 16 (melanoma differentiation); IL-24; IL-4-induced secreted protein; Interleukin-24; MDA-7; Melanoma differentiation-associated gene 7 protein; Th2-specific cytokine FISP
Gene Aliases: FISP; Il24; Mda-7; Mda7; St16
UniProt ID: (Mouse) Q925S4
Entrez Gene ID: (Mouse) 93672
Molecular Function: cytokine interleukin superfamily signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.