Invitrogen
Manufacturer Code:PA576670
Catalog # PIPA576670
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human IL-24. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C49AFISPIL 24IL 4 induced secreted proteinIL-24IL10BIl24IL24&lowbarHUMANinterleukin 24Interleukin-24MDA-7MDA7Melanocyte associated Mda 7Melanoma differentiation association protein 7Melanoma differentiation-associated gene 7 proteinMob5ST16Suppression of tumorigenicity 16 (melanoma differentiation)Suppression of tumorigenicity 16Suppression of tumorigenicity 16 protein; IL-24; IL-4-induced secreted protein; Interleukin-24; MDA-7; melanocyte-associated Mda-7; Melanoma differentiation-associated gene 7 protein; suppression of tumorigenicity 16 (melanoma differentiation); Suppression of tumorigenicity 16 protein; Th2-specific cytokine FISP
Gene Aliases: C49A; FISP; IL10B; IL24; Mda-7; MDA7; MOB5; ST16
UniProt ID: (Human) Q13007, (Mouse) Q925S4
Entrez Gene ID: (Human) 11009, (Mouse) 93672
Molecular Function:
cytokine
interleukin superfamily
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.