Invitrogen
Manufacturer Code:PA5102507
Catalog # PIPA5102507
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human IL24 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C49A Cytokine-like protein Mob-5 FISP IL 24 IL10B Il24 IL-24 IL-4-induced secreted protein interleukin 24 interleukin-24 MDA7 MDA-7 melanocyte-associated Mda-7 melanoma differentiation-associated gene 7 protein Mob5 St16 suppression of tumorigenicity 16 (melanoma differentiation) Suppression of tumorigenicity 16 protein Th2-specific cytokine FISP; Cytokine-like protein Mob-5; IL-24; IL-4-induced secreted protein; Interleukin-24; MDA-7; melanocyte-associated Mda-7; Melanoma differentiation-associated gene 7 protein; suppression of tumorigenicity 16 (melanoma differentiation); Suppression of tumorigenicity 16 protein; Th2-specific cytokine FISP
Gene Aliases: C49A; FISP; IL10B; IL24; Mda-7; MDA7; MOB5; ST16
UniProt ID: (Human) Q13007, (Rat) Q9JI24, (Mouse) Q925S4
Entrez Gene ID: (Human) 11009, (Rat) 170819, (Mouse) 93672
Molecular Function: cytokine interleukin superfamily signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.