Invitrogen
Manufacturer Code:MA523906
Catalog # PIMA523906
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 258010 |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse IL-23 R Gly24-Asp372 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: IBD17 IL-23 receptor IL-23R interleukin 23 receptor interleukin-23 receptor; IL-23 receptor; Interleukin-23 receptor
Gene Aliases: IL-23R; Il23r
UniProt ID: (Mouse) Q8K4B4
Entrez Gene ID: (Mouse) 209590
Molecular Function: cytokine defense/immunity protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.