Invitrogen
Manufacturer Code:PA579490
Catalog # PIPA579490
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived mouse IL23 Receptor recombit protein (Position: I25-D233). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: IL-23 receptor; interleukin 23 receptor; interleukin 23 receptor-like; Interleukin-23 receptor; Interleukin-23 receptor IL-23 receptor IL-23R Il23r
Gene Aliases: IL-23R; Il23r
UniProt ID: (Mouse) Q8K4B4
Entrez Gene ID: (Rat) 689932, (Mouse) 209590
Molecular Function: cytokine defense/immunity protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.