Invitrogen
Manufacturer Code:PA5113004
Catalog # PIPA5113004
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Interleukin-23 receptor protein (15-125AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: IBD17 IL 23R IL-23 receptor Il23r IL-23R interleukin 23 receptor interleukin 23 receptor-like Interleukin-23 receptor; IL-23 receptor; Interleukin-23 receptor
Gene Aliases: IL-23R; IL23R
UniProt ID: (Human) Q5VWK5, (Mouse) Q8K4B4
Entrez Gene ID: (Human) 149233, (Mouse) 209590
Molecular Function: cytokine defense/immunity protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.