Invitrogen
Manufacturer Code:PA587317
Catalog # PIPA587317
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human IL21R. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD360; IL-21 receptor; IL-21R; Interleukin-21 receptor; Interleukin-21 receptor IL-21 receptor IL-21R Novel interleukin receptor CD360 IL21R NILR UNQ3121/PRO10273; LR-beta; Lymphocyte receptor beta; Novel cytokine receptor 8; Novel interleukin receptor; NR8
Gene Aliases: CD360; IL21R; NILR; UNQ3121/PRO10273
UniProt ID: (Human) Q9HBE5, (Mouse) Q9JHX3
Entrez Gene ID: (Human) 50615, (Rat) 308977, (Mouse) 60504
Molecular Function: cytokine receptor receptor type I cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.