Invitrogen
Manufacturer Code:PA129843
Catalog # PIPA129843
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides (CSLHRSAHNATHATYT & CDFTSPGDEGPPRSYLR) corresponding to amino acids 65-80 and 505-521 of Human IL21R respectively. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD360; IL-21 receptor; IL-21R; Interleukin-21 receptor; LR-beta; Lymphocyte receptor beta; Novel cytokine receptor 8; Novel interleukin receptor; NR8
Gene Aliases: CD360; IL21R; NILR; UNQ3121/PRO10273
UniProt ID: (Human) Q9HBE5, (Mouse) Q9JHX3
Entrez Gene ID: (Human) 50615, (Mouse) 60504
Molecular Function:
cytokine receptor
receptor
type I cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.