Abnova
Manufacturer Code:PAB17987
Catalog # 89-119-593
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 20-32 of human IL21R. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of IL21R.
For Research Use Only
Protein Aliases: CD360; IL-21 receptor; Interleukin-21 receptor; MGC10967; NILR; Novel interleukin receptor
Gene Aliases: CD360; IL21R; NILR; UNQ3121/PRO10273
UniProt ID: (Human) Q9HBE5
Entrez Gene ID: (Human) 50615
Molecular Function:
cytokine receptor
receptor
type I cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.