Invitrogen
Manufacturer Code:PA538013
Catalog # PIPA538013
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | 16 amino acid peptide near the center of human IL-1RL2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: IL-1 receptor-related protein 2; IL-1Rrp2; IL-36 receptor; IL-36R; IL-36R IL1RRP2 IL-1Rrp2 IL1R-rp2 Interleukin-1 receptor-like 2 IL-36 receptor IL-36R; IL1R-rp2; interleukin 1 receptor-like 2; interleukin-1 receptor related protein 2; Interleukin-1 receptor-like 2; Interleukin-1 receptor-related protein 2
Gene Aliases: AI481289; AW551444; IL-1Rrp2; IL-36R; IL1R-rp2; IL1RL2; IL1RRP2
UniProt ID: (Human) Q9HB29, (Rat) Q62929, (Mouse) Q9ERS7
Entrez Gene ID: (Human) 8808, (Rat) 171106, (Mouse) 107527
Molecular Function: cytokine receptor receptor type I cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.