Proteintech
Manufacturer Code:216091AP150UL
Catalog # 50-173-3511
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | IL1RAPL1 Fusion Protein Ag16252 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: IL 1 RAPL 1 IL 1RAPL 1 IL1R8 IL1RAPL IL1RAPL 1 IL1RAPL1 MRX10 MRX21 MRX34 Oligophrenin 4 OPHN4 TIGIRR 2; IL-1-RAPL-1; IL-1RAPL-1; IL1RAPL-1; interleukin 1 receptor accessory protein-like 1; interleukin 1 receptor-8; Interleukin-1 receptor accessory protein-like 1; mental retardation, X-linked 10; Oligophrenin-4; Three immunoglobulin domain-containing IL-1 receptor-related 2; TIGIRR-2; X-linked interleukin-1 receptor accessory protein-like 1
Gene Aliases: 6330532G10Rik; C030039N24; IL1R8; IL1RAPL; IL1RAPL1; MRX10; MRX21; MRX34; OPHN4; TIGIRR-2
UniProt ID: (Human) Q9NZN1, (Rat) P59824
Entrez Gene ID: (Human) 11141, (Mouse) 331461, (Rat) 317553
Molecular Function:
cytokine receptor
receptor
type I cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.