Invitrogen
Manufacturer Code:PA547661
Catalog # PIPA547661
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived recombit mouse IL-1ra/IL-1F3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: IL-1RN; IL1 inhibitor; Interleukin-1 receptor antagonist protein; Interleukin-1 receptor antagonist protein IL-1RN IL-1ra IRAP ICIL-1RA IL1 inhibitor Anakinra IL1RN IL1F3 IL1RA IL1-RA; IRAP
Gene Aliases: F630041P17Rik; IL-1ra; Il1rn
UniProt ID: (Mouse) P25085
Entrez Gene ID: (Mouse) 16181
Molecular Function: cytokine interleukin superfamily signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.