Invitrogen
Manufacturer Code:PA586880
Catalog # PIPA586880
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to residues in Human IL1R2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: antigen CDw121b; CD121 antigen-like family member B; CD121b; CDw121b; IL-1 receptor beta chain; IL-1 type II receptor; IL-1R-2; IL-1R-beta; IL-1RT-2; IL-1RT2; interleukin 1 receptor type II variant 3; interleukin 1 receptor, type II; Interleukin-1 receptor beta; Interleukin-1 receptor type 2; Interleukin-1 receptor type 2 IL-1R-2 IL-1RT-2 IL-1RT2 CD121 antigen-like family member B CDw121b IL-1 type II receptor Interleukin-1 receptor beta IL-1R-beta Interleukin-1 receptor type II CD121b Interleukin-1 receptor type 2 membrane form mIL-1R2 mIL-1RII Interleukin-1 receptor type 2 soluble form sIL-1R2 sIL-1RII IL1R2 IL1RB; Interleukin-1 receptor type 2, membrane form; Interleukin-1 receptor type 2, soluble form; Interleukin-1 receptor type II; mIL-1R2; sIL-1R2; type II interleukin-1 receptor, beta
Gene Aliases: CD121b; CDw121b; IL-1R-2; Il-1r2; IL-1RT-2; IL-1RT2; Il1r-2; IL1R2; IL1R2c; IL1RB
UniProt ID: (Human) P27930, (Mouse) P27931
Entrez Gene ID: (Human) 7850, (Mouse) 16178
Molecular Function: cytokine receptor receptor type I cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.